Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205026.1 | NP_990357.1 | 8 - 9 | 680 | 80 | |
XM_015296335.2 | XP_015151821.1 | 8 - 9 | 700 | 80 | ||
XM_025142176.1 | XP_024997944.1 | 8 - 9 | 707 | 80 | ||
GenBank mRNA | AB219158.1 | - | 7 - 8 | 634 | 80 | |
AF029841.1 | - | 8 - 9 | 680 | 80 | ||
HG007958.1 | - | 8 - 9 | 713 | 80 | ||
LC144559.1 | - | 7 - 8 | 634 | 80 | ||
LC144560.1 | - | 7 - 8 | 634 | 80 | ||
LC144561.1 | - | 7 - 8 | 634 | 80 | ||
LC144563.1 | - | 7 - 8 | 634 | 80 | ||
LC144566.1 | - | 7 - 8 | 634 | 80 | ||
LC144571.1 | - | 7 - 8 | 634 | 80 | ||
LC144572.1 | - | 7 - 8 | 634 | 80 | ||
LC144573.1 | - | 7 - 8 | 634 | 80 | ||
LC144577.1 | - | 7 - 8 | 634 | 80 | ||
LC144578.1 | - | 7 - 8 | 634 | 80 | ||
LC144579.1 | - | 7 - 8 | 634 | 80 | ||
LC144580.1 | - | 7 - 8 | 634 | 80 | ||
LC144581.1 | - | 7 - 8 | 634 | 80 | ||
LC144583.1 | - | 7 - 8 | 634 | 80 | ||
LC144584.1 | - | 7 - 8 | 634 | 80 | ||
LC144585.1 | - | 7 - 8 | 634 | 80 | ||
LC144587.1 | - | 7 - 8 | 634 | 80 | ||
LC144588.1 | - | 7 - 8 | 634 | 80 | ||
LC144589.1 | - | 7 - 8 | 634 | 80 | ||
LC144590.1 | - | 7 - 8 | 634 | 80 | ||
LC144591.1 | - | 7 - 8 | 634 | 80 | ||
LC144592.1 | - | 7 - 8 | 634 | 80 | ||
LC144593.1 | - | 7 - 8 | 634 | 80 | ||
LC144594.1 | - | 7 - 8 | 634 | 80 | ||
LC144595.1 | - | 7 - 8 | 634 | 80 | ||
LC144596.1 | - | 7 - 8 | 634 | 80 | ||
LC144597.1 | - | 7 - 8 | 634 | 80 | ||
LC144598.1 | - | 7 - 8 | 634 | 80 | ||
LC144599.1 | - | 7 - 8 | 634 | 80 | ||
LC144600.1 | - | 7 - 8 | 634 | 80 | ||
LC144601.1 | - | 7 - 8 | 634 | 80 | ||
LC144603.1 | - | 7 - 8 | 634 | 80 | ||
LC144604.1 | - | 7 - 8 | 634 | 80 | ||
LC144605.1 | - | 7 - 8 | 634 | 80 | ||
LC144607.1 | - | 7 - 8 | 634 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |