Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205027.1 | NP_990358.1 | 6 - 7 | 735 | 71 | |
XM_015280589.2 | XP_015136075.1 | 7 - 8 | 797 | 71 | ||
XM_015280591.2 | XP_015136077.1 | 7 - 8 | 1679 | 71 | ||
XM_025144671.1 | XP_025000439.1 | 6 - 7 | 1527 | 71 | ||
GenBank mRNA | AB052132.1 | - | 6 - 7 | 686 | 71 | |
L36678.1 | - | 6 - 7 | 735 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |