Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205237.3 | NP_990568.3 | 2 - 3 | 502 | 106 | |
GenBank mRNA | BM426853.1 | - | 2 - 3 | 487 | 106 | |
MH553645.1 | - | 1 - 2 | 372 | 106 | ||
MH553646.1 | - | 1 - 2 | 372 | 106 | ||
X75915.1 | - | 375 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |