Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190165.4 | NP_001177094.1 | 4 - 5 | 413 | 94 | |
NM_205260.5 | NP_990591.1 | 4 - 5 | 413 | 94 | ||
NR_073182.3 | 4 - 5 | 402 | 94 | |||
XM_015289639.2 | XP_015145125.1 | 5 - 6 | 4184 | 94 | ||
GenBank mRNA | DQ185930.1 | - | 4 - 5 | 413 | 94 | |
DQ185931.1 | - | 4 - 5 | 413 | 94 | ||
S78477.1 | - | 4 - 5 | 411 | 94 | ||
Y07539.1 | - | 403 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |