Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205363.1 | NP_990694.1 | 8 - 9 | 906 | 100 | |
XM_015288596.2 | XP_015144082.1 | 8 - 9 | 966 | 100 | ||
GenBank mRNA | L38519.1 | - | 8 - 9 | 906 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |