Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205473.1 | NP_990804.1 | 2 - 3 | 220 | 84 | |
GenBank mRNA | M87294.1 | - | 2 - 3 | 220 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |