Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030347.1 | NP_001025518.1 | 3 - 4 | 346 | 80 | |
GenBank mRNA | AF035677.1 | - | 3 - 4 | 346 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |