Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030385.1 | NP_001025556.1 | 5 - 6 | 1060 | 81 | |
XM_015278797.2 | XP_015134283.1 | 4 - 5 | 1430 | 81 | ||
XM_015278798.2 | XP_015134284.1 | 5 - 6 | 1151 | 81 | ||
GenBank mRNA | AJ719662.1 | - | 5 - 6 | 1060 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |