Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030562.2 | NP_001025733.2 | 5 - 6 | 408 | 128 | |
NM_001305547.1 | NP_001292476.1 | 4 - 5 | 369 | 128 | ||
NM_001305548.1 | NP_001292477.1 | 4 - 5 | 504 | 128 | ||
XM_015279126.2 | XP_015134612.1 | 4 - 5 | 360 | 128 | ||
GenBank mRNA | AJ719600.1 | - | 5 - 6 | 409 | 128 | |
BU116275.1 | - | 4 - 5 | 369 | 128 | ||
BU144410.1 | - | 4 - 5 | 355 | 128 | ||
BU345205.1 | - | 4 - 5 | 504 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |