Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF4beta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030576.4 | NP_001025747.3 | 5 - 6 | 726 | 90 | |
GenBank mRNA | AY700582.1 | - | 5 - 6 | 830 | 90 | |
BU300422.1 | - | 3 - 4 | 354 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |