Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030609.2 | NP_001025780.2 | 1 - 2 | 247 | 119 | |
NM_001305576.1 | NP_001292505.1 | 1 - 2 | 247 | 119 | ||
GenBank mRNA | AJ720113.1 | - | 1 - 2 | 232 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |