Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030611.2 | NP_001025782.2 | 2 - 3 | 342 | 101 | |
GenBank mRNA | AJ720936.1 | - | 2 - 3 | 364 | 101 | |
CR733192.1 | - | 2 - 3 | 328 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |