Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030802.1 | NP_001025973.1 | 3 - 4 | 1532 | 135 | |
XM_015302426.2 | XP_015157912.1 | 4 - 5 | 1868 | 135 | ||
GenBank mRNA | AJ720342.1 | - | 3 - 4 | 1532 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |