Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030841.2 | NP_001026012.1 | 2 - 3 | 514 | 88 | |
GenBank mRNA | AJ851411.1 | - | 2 - 3 | 517 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |