Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030841.2 | NP_001026012.1 | 4 - 5 | 2423 | 77 | |
XM_015293603.2 | XP_015149089.1 | 3 - 4 | 9513 | 77 | ||
GenBank mRNA | AJ851411.1 | - | 4 - 5 | 2426 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |