Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030942.1 | NP_001026113.1 | 4 - 5 | 572 | 127 | |
XM_015282807.2 | XP_015138293.2 | 4 - 5 | 474 | 127 | ||
GenBank mRNA | AJ720953.1 | - | 4 - 5 | 572 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |