Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031018.1 | NP_001026189.1 | 3 - 4 | 331 | 107 | |
XM_015282591.2 | XP_015138077.1 | 4 - 5 | 432 | 107 | ||
XM_025147306.1 | XP_025003074.1 | 3 - 4 | 405 | 107 | ||
GenBank mRNA | AJ851507.1 | - | 3 - 4 | 331 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |