Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031026.1 | NP_001026197.1 | 156 | 105 | ||
XM_015298703.2 | XP_015154189.1 | 172 | 105 | |||
XM_025143709.1 | XP_024999477.1 | 172 | 105 | |||
GenBank mRNA | AJ720479.1 | - | 156 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus |