Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031026.1 | NP_001026197.1 | 459 | 126 | ||
XM_015298703.2 | XP_015154189.1 | 475 | 126 | |||
GenBank mRNA | AJ720479.1 | - | 459 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus |