Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031040.1 | NP_001026211.1 | 5 - 6 | 1343 | 74 | |
GenBank mRNA | AJ719567.1 | - | 5 - 6 | 1343 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |