Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031227.1 | NP_001026398.1 | 6 - 7 | 1088 | 135 | |
XM_015288218.2 | XP_015143704.1 | 7 - 8 | 988 | 135 | ||
GenBank mRNA | AJ719337.1 | - | 6 - 7 | 1088 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |