Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031227.1 | NP_001026398.1 | 8 - 9 | 1432 | 95 | |
XM_015288218.2 | XP_015143704.1 | 9 - 10 | 1332 | 95 | ||
GenBank mRNA | AJ719337.1 | - | 8 - 9 | 1432 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |