Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031238.1 | NP_001026409.1 | 7 - 8 | 1099 | 131 | |
GenBank mRNA | AJ719355.1 | - | 7 - 8 | 1099 | 131 | |
CR353948.1 | - | 5 - 6 | 669 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |