Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTPCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031523.1 | NP_001026694.1 | 2 - 3 | 216 | 142 | |
GenBank mRNA | AJ719769.1 | - | 2 - 3 | 216 | 142 | |
KT153608.1 | - | 2 - 3 | 193 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |