Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031585.2 | NP_001026756.1 | 3 - 4 | 1638 | 139 | |
XM_015283175.2 | XP_015138661.1 | 3 - 4 | 1704 | 139 | ||
GenBank mRNA | AJ851559.1 | - | 3 - 4 | 1639 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |