Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031597.1 | NP_001026768.1 | 8 - 9 | 1719 | 88 | |
XM_025147342.1 | XP_025003110.1 | 8 - 9 | 1786 | 88 | ||
GenBank mRNA | AJ719881.1 | - | 8 - 9 | 1719 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |