Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030338.1 | NP_001025509.1 | 3 - 4 | 1352 | 88 | |
GenBank mRNA | AJ238354.1 | - | 3 - 4 | 1352 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |