Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030652.1 | NP_001025823.1 | 2 - 3 | 449 | 92 | |
XM_015275227.2 | XP_015130713.1 | 2 - 3 | 464 | 92 | ||
GenBank mRNA | AJ719634.1 | - | 2 - 3 | 449 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |