Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270716.1 | NP_001257645.1 | 4 - 5 | 327 | 116 | |
XM_015275319.2 | XP_015130805.1 | 4 - 5 | 332 | 116 | ||
GenBank mRNA | AJ721085.1 | - | 3 - 4 | 704 | 116 | |
BU296761.1 | - | 4 - 5 | 337 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |