Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030681.1 | NP_001025852.1 | 2 - 3 | 1102 | 74 | |
GenBank mRNA | AJ720914.1 | - | 2 - 3 | 1102 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |