Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030711.1 | NP_001025882.1 | 6 - 7 | 939 | 93 | |
XM_015295722.2 | XP_015151208.1 | 6 - 7 | 944 | 93 | ||
XM_015295723.2 | XP_015151209.1 | 6 - 7 | 1004 | 93 | ||
XM_015295725.2 | XP_015151211.1 | 6 - 7 | 1015 | 93 | ||
XM_025141948.1 | XP_024997716.1 | 7 - 8 | 2229 | 93 | ||
XM_025141949.1 | XP_024997717.1 | 5 - 6 | 852 | 93 | ||
XM_025141950.1 | XP_024997718.1 | 5 - 6 | 950 | 93 | ||
GenBank mRNA | AJ720958.1 | - | 6 - 7 | 939 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |