Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030726.1 | NP_001025897.1 | 6 - 7 | 1042 | 123 | |
XM_015295802.2 | XP_015151288.2 | 6 - 7 | 858 | 123 | ||
XM_015295804.2 | XP_015151290.2 | 5 - 6 | 830 | 123 | ||
GenBank mRNA | AJ851702.1 | - | 6 - 7 | 1042 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |