Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030763.2 | NP_001025934.2 | 4 - 5 | 583 | 112 | |
XM_025148736.1 | XP_025004504.1 | 4 - 5 | 576 | 112 | ||
XM_025148737.1 | XP_025004505.1 | 4 - 5 | 586 | 112 | ||
GenBank mRNA | AJ720065.1 | - | 4 - 5 | 583 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |