Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030790.1 | NP_001025961.1 | 1 - 2 | 156 | 57 | |
NM_001278101.3 | NP_001265030.1 | 1 - 2 | 173 | 57 | ||
GenBank mRNA | AJ719393.1 | - | 1 - 2 | 156 | 57 | |
BI066001.1 | - | 1 - 2 | 160 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |