Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030894.2 | NP_001026065.1 | 5 - 6 | 608 | 116 | |
XM_015297648.2 | XP_015153134.1 | 6 - 7 | 755 | 116 | ||
GenBank mRNA | AJ719961.1 | - | 5 - 6 | 609 | 116 | |
BX929636.1 | - | 1 - 2 | 621 | 116 | ||
BX930386.1 | - | 5 - 6 | 591 | 116 | ||
BX930954.1 | - | 6 - 7 | 685 | 116 | ||
BX932588.1 | - | 5 - 6 | 592 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |