Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBA57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030958.2 | NP_001026129.2 | 2 - 3 | 684 | 87 | |
XM_015281082.2 | XP_015136568.1 | 2 - 3 | 423 | 87 | ||
XM_015281083.2 | XP_015136569.1 | 2 - 3 | 401 | 87 | ||
GenBank mRNA | AJ719942.1 | - | 2 - 3 | 684 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |