Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030969.1 | NP_001026140.1 | 1 - 2 | 452 | 88 | |
GenBank mRNA | AJ720905.1 | - | 1 - 2 | 452 | 88 | |
KY747396.1 | - | 1 - 2 | 130 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |