Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030986.1 | NP_001026157.1 | 4 - 5 | 330 | 85 | |
XM_015281467.1 | XP_015136953.1 | 3 - 4 | 364 | 85 | ||
XM_015281468.2 | XP_015136954.1 | 3 - 4 | 234 | 85 | ||
XM_025147204.1 | XP_025002972.1 | 4 - 5 | 360 | 85 | ||
GenBank mRNA | AJ719665.1 | - | 4 - 5 | 391 | 85 | |
AJ720825.1 | - | 4 - 5 | 330 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |