Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031310.1 | NP_001026481.1 | 4 - 5 | 363 | 109 | |
GenBank mRNA | AJ720946.1 | - | 4 - 5 | 363 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |