Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031335.1 | NP_001026506.1 | 4 - 5 | 392 | 58 | |
GenBank mRNA | AJ720278.1 | - | 4 - 5 | 392 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |