Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031341.1 | NP_001026512.1 | 5 - 6 | 867 | 70 | |
XM_025143981.1 | XP_024999749.1 | 4 - 5 | 640 | 70 | ||
GenBank mRNA | AJ720268.1 | - | 5 - 6 | 867 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |