Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031370.2 | NP_001026541.1 | 4 - 5 | 1014 | 102 | |
GenBank mRNA | AJ851535.1 | - | 4 - 5 | 1015 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |