Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNSTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031393.1 | NP_001026564.1 | 3 - 4 | 337 | 69 | |
GenBank mRNA | AJ721023.1 | - | 3 - 4 | 337 | 69 | |
CR352610.1 | - | 2 - 3 | 191 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |