Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031394.1 | NP_001026565.1 | 2 - 3 | 395 | 94 | |
GenBank mRNA | AJ720171.1 | - | 2 - 3 | 395 | 94 | |
CR352406.1 | - | 2 - 3 | 326 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |