Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031409.1 | NP_001026580.1 | 12 - 13 | 1227 | 101 | |
XM_015293130.2 | XP_015148616.1 | 13 - 14 | 1472 | 101 | ||
XM_015293131.2 | XP_015148617.1 | 13 - 14 | 1344 | 101 | ||
XM_015293132.2 | XP_015148618.1 | 12 - 13 | 1405 | 101 | ||
GenBank mRNA | AJ868233.1 | - | 12 - 13 | 1227 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |