Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031409.1 | NP_001026580.1 | 3 - 4 | 427 | 89 | |
XM_015293130.2 | XP_015148616.1 | 4 - 5 | 672 | 89 | ||
XM_015293131.2 | XP_015148617.1 | 4 - 5 | 544 | 89 | ||
XM_015293132.2 | XP_015148618.1 | 4 - 5 | 674 | 89 | ||
GenBank mRNA | AJ868233.1 | - | 3 - 4 | 427 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |