Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033850.1 | NP_001029022.1 | 7 - 8 | 764 | 72 | |
NM_001193588.1 | NP_001180517.1 | 9 - 10 | 911 | 72 | ||
NM_001193589.1 | NP_001180518.1 | 8 - 9 | 836 | 72 | ||
XM_025150837.1 | XP_025006605.1 | 9 - 10 | 1094 | 72 | ||
GenBank mRNA | AB234613.1 | - | 9 - 10 | 911 | 72 | |
AB234614.1 | - | 8 - 9 | 836 | 72 | ||
AB234615.1 | - | 7 - 8 | 764 | 72 | ||
AB376101.1 | - | 2 - 3 | 269 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |