Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034814.2 | NP_001029986.2 | 1 - 2 | 170 | 64 | |
XM_025154152.1 | XP_025009920.1 | 8 - 9 | 4454 | 64 | ||
GenBank mRNA | AB205527.1 | - | 1 - 2 | 196 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |