Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039255.1 | NP_001034344.1 | 1 - 2 | 170 | 60 | |
GenBank mRNA | AJ720244.1 | - | 1 - 2 | 170 | 60 | |
BX931383.1 | - | 165 | 60 | |||
BX932666.2 | - | 1 - 2 | 166 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |