Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039255.1 | NP_001034344.1 | 6 - 7 | 705 | 96 | |
GenBank mRNA | AJ720244.1 | - | 6 - 7 | 705 | 96 | |
BX932666.2 | - | 6 - 7 | 701 | 96 | ||
KU325499.1 | - | 5 - 6 | 524 | 96 | ||
KU557484.1 | - | 5 - 6 | 524 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |